Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAMHD1 Antibody (OTI3F5), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273979B
Description
SAMHD1 Monoclonal specifically detects SAMHD1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
SAMHD1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SAMHD1 | |
Full length recombinant protein of human SAMHD1 (NP_056289) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3F5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DCIPMg11, Dendritic cell-derived IFNG-induced protein, EC 3.1.4.-, HDDC1, Monocyte protein 5, MOP5, MOP-5AGS5, SAM domain and HD domain 1, SAM domain and HD domain-containing protein 1, SBBI88 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
25939.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction