Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAMHD1 Antibody (OTI3F5), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP203285
Description
SAMHD1 Monoclonal specifically detects SAMHD1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SAMHD1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:200-500, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
SAMHD1 | |
Full length human recombinant protein of human SAMHD1(NP_056289) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI3F5 | |
Unconjugated | |
DCIPMg11, Dendritic cell-derived IFNG-induced protein, EC 3.1.4.-, HDDC1, Monocyte protein 5, MOP5, MOP-5AGS5, SAM domain and HD domain 1, SAM domain and HD domain-containing protein 1, SBBI88 | |
Mouse | |
72 kDa | |
0.1 mL | |
Stem Cell Markers | |
25939 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction