Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAMHD1 Monoclonal Antibody (OTI1F9), TrueMAB™, OriGene
SDP

Catalog No. 502541896
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1896 30 μL
50-167-3051 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1896 Supplier OriGene Supplier No. TA501955S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SAMHD1 Monoclonal antibody specifically detects SAMHD1 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen SAMHD1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1F9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SAMHD1
Gene Accession No. Q9Y3Z3
Gene Alias CHBL2; DCIP; Dendritic cell-derived IFNG-induced protein; deoxynucleoside triphosphate triphosphohydrolase SAMHD1; dNTPase; E330031J07Rik; EGK_02438; HDDC1; hSAMHD1; IFN-gamma induced; interferon-gamma-inducible protein Mg11; Mg11; Mg21; Monocyte protein 5; MOP5; MOP-5; mSAMHD1; RP1-132F21.1; SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1; SAM domain and HD domain 1; SAM domain and HD domain, 1; SAM domain and HD domain-containing protein 1; Samhd1; SBBI88
Host Species Mouse
Immunogen Full length human recombinant protein of human SAMHD1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25939
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.