Learn More
SAMHD1 Mouse anti-Human, Clone: OTI3G6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502025

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene may play a role in regulation of the innate immune response. The encoded protein is upregulated in response to viral infection and may be involved in mediation of tumor necrosis factor-alpha proinflammatory responses. Mutations in this gene have been associated with Aicardi-Goutieres syndrome.Specifications
SAMHD1 | |
Monoclonal | |
Unconjugated | |
SAMHD1 | |
CHBL2, DCIP, HDDC1, MOP-5, SBBI88 | |
Mouse | |
Affinity Chromatography | |
RUO | |
25939 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3G6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y3Z3 | |
SAMHD1 | |
Full length human recombit protein of human SAMHD1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.