Learn More
SAMHD1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303690AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
This gene may play a role in regulation of the innate immune response. The encoded protein is upregulated in response to viral infection and may be involved in mediation of tumor necrosis factor-alpha proinflammatory responses. Mutations in this gene have been associated with Aicardi-Goutieres syndrome.Specifications
SAMHD1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9Y3Z3 | |
SAMHD1 | |
Between 450 and 500 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
SAMHD1 | |
RP1-132F21.1, CHBL2, DCIP, HDDC1, MOP-5, SBBI88, SAM domain and HD domain-containing protein 1, dendritic cell-derived IFNG-induced protein, monocyte protein 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25939 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.