Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAMM50 Antibody (SR1928), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322489100UL
Description
SAMM50 Monoclonal antibody specifically detects SAMM50 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
SAMM50 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1928 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
CGI-51, FLJ42905, FLJ99036, OMP85, SAM50, sorting and assembly machinery 50kDa, sorting and assembly machinery component 50 homolog, sorting and assembly machinery component 50 homolog (S. cerevisiae), TOB55, Transformation-related gene 3 protein, transformation-related protein 3, TRG-3FLJ35825, YNL026W | |
A synthesized peptide derived from human SAMM50 (Uniprot #: Q9Y512) | |
100 μg | |
Stem Cell Markers | |
25813 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction