Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Sample Diluent Concentrate 2 (2X)
SDP

Catalog No. DYC002 Shop All Novus Biologicals Products
This item is not returnable. View return policy
Catalog No. DYC002 Supplier R&D Systems™ Supplier No. DYC002

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains 2% NP-40 Alternative, 40mM Tris, 274mM NaCl, 20% glycerol, 2mM EDTA, and 2mM activated sodium orthovanadate

Specifications

For Use With (Application) ELISA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.