Learn More
IKA™ Sample Flask Bottle 1000 mL
SDP
Catalog No. 11116948 Shop All IKA Works Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
Catalog No. Capacity (Metric)
11-116-948 1000 mL
1 options
Catalog No. 11-116-948 Supplier IKA™ Supplier No. 0020110853
Add to Cart
Add to Cart
This 1000 mL Sample bottle can be used to feed various liquids into the Habitat bioreactor (e.g. acid, base, antifoam agents or nutrient solutions).
  • Designed for use with IKA Habitat bioreactor
  • 1000 mL volume
Specifications
Capacity (Metric) 1000 mL