Learn More
IKA™ Sample Flask Bottle
Designed to feed various liquids into the Habitat bioreactor
Supplier: IKA™ 0020110852
Description
- Sample bottle with a volume of 500 mL
- This can be used to feed various liquids into the Habitat bioreactor (e.g. acid, base, antifoam agents or nutrient solutions)
- Designed for use with IKA Habitat bioreactor
- 500 mL volume
Specifications
500 mL |