Learn More
IKA™ Sample Flask Rack
SDP
Catalog No. 11116949 Shop All IKA Works Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Catalog No. Capacity (Metric)
11-116-949 250 mL
1 options
Catalog No. 11-116-949 Supplier IKA™ Supplier No. 0020110862
Add to Cart
Add to Cart
With the help of this stand, you can safely store your sample bottles. There are 4 positions for 250 mL bottles. Due to a slightly inclined position, the sample bottle can be completely emptied without any problems.
  • 4 positions for 250 mL bottles
  • Inclined position
Specifications
Capacity (Metric) 250 mL
Material Glass