Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPEX™ SamplePrep DNAmax Isolation Kit
Click to view available options
Sample Type:
Animal Tissue
Soft Plant Tissue
Tough Plant Tissue
Buffer:
Lysis, Wash, Elution
Description
- The complete kit solution for DNA extraction of animal and plant tissue
- Reinforced tubes designed for homogenization through rapid beat beating
- Silica-based columns capture DNA and allow for its purification
- Protocol and workflow optimized for Genomax, Geno/Grinder, and GenoLyte
Specifications
Specifications
Buffer | Lysis, Wash, Elution |
Bead Type | Stainless Steel |
Sample Type | Animal Tissue |
For Use With (Equipment) | Genomax, Geno/Grinder, GenoLyte |
For Use With (Application) | DNA Extraction of Animal and Plant Tissue |
Includes | Lysis Buffer, Wash Buffer, Elution Buffer, Proteinase K Powder, Rnase A, 3 mm Stainless Steel Balls |
No. of Reactions | 75 |
Product Type | DNA Isolation Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction