Learn More
SAMSN1 Mouse anti-Human, Clone: OTI7C12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808901

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SAMSN (HACS1) is a member of a novel gene family of putative adaptors and scaffold proteins containing SH3 and SAM (sterile alpha motif) domains. It encodes a 441 amino acid protein that is differentially expressed in hematopoietic cells and maligcies including myeloid leukemia, lymphoma, and myeloma. SAMSN has restricted expression in human tissues. It has been shown to be up-regulated by B cell activation signals and is a participant in B cell activation and differentiation. SAMSN is an immunoinhibitory adaptor that might be a useful target for immune suppression therapy.Specifications
SAMSN1 | |
Monoclonal | |
Unconjugated | |
SAMSN1 | |
HACS1, NASH1, SASH2, SH3D6B, SLy2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
64092 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI7C12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NSI8 | |
SAMSN1 | |
Human recombit protein fragment corresponding to amino acids 1-250 of human SAMSN1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.