Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAMSN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582400 Supplier Invitrogen™ Supplier No. PA582400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TEAHEGDPTN GSGEQSKTSN NGGGLGKKMR AISWTMKKKV GKKYIKALSE EKDEEDGENA HPYRNSDPVI GTHTEKVSLK ASDSMDSLYS GQSSSSGITS CSDGTSNRDS FRLDDDGPYS GPFC Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 83%.

SAMSN (HACS1) is a member of a novel gene family of putative adaptors and scaffold proteins containing SH3 and SAM (sterile alpha motif) domains. It encodes a 441 amino acid protein that is differentially expressed in hematopoietic cells and malignancies including myeloid leukemia, lymphoma, and myeloma. SAMSN has restricted expression in human tissues. It has been shown to be up-regulated by B cell activation signals and is a participant in B cell activation and differentiation. SAMSN is an immunoinhibitory adaptor that might be a useful target for immune suppression therapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAMSN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAMSN1
Gene Accession No. Q9NSI8
Gene Alias 4930571B16Rik; 930571B16Rik; HACS1; hematopoietic adapter-containing SH3 and sterile α-motif (SAM) domains 1; hematopoietic adapter-containing SH3 and sterile alpha-motif (SAM) domains 1; hematopoietic adaptor containing SH3 and SAM domains 1; Nash; NASH1; nuclear localization signals, SAM and SH3 domain containing 1; SAM and SH3 domain containing 2; SAM domain, SH3 domain and nuclear localisation signals, 1; SAM domain, SH3 domain and nuclear localization signals 1; SAM domain, SH3 domain and nuclear localization signals protein 1; SAM domain, SH3 domain and nuclear localization signals, 1; SAM domain-containing protein SAMSN-1; SAMSN; Samsn1; SAMSN-1; SASH2; SH3 protein expressed in lymphocytes 2; SH3D6B; SH3-lymphocyte protein 2; SH3-SAM adaptor protein; SLy2; Src homology domain 3 (SH3)-containing adapter protein SH3 lymphocyte protein 2
Gene Symbols SAMSN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAMSN1. Recombinant protein control fragment (Product #RP-89850).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64092
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.