Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAMSN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100326
Description
Antibody detects endogenous levels of total SAMSN1.
SAMSN (HACS1) is a member of a novel gene family of putative adaptors and scaffold proteins containing SH3 and SAM (sterile alpha motif) domains. It encodes a 441 amino acid protein that is differentially expressed in hematopoietic cells and malignancies including myeloid leukemia, lymphoma, and myeloma. SAMSN has restricted expression in human tissues. It has been shown to be up-regulated by B cell activation signals and is a participant in B cell activation and differentiation. SAMSN is an immunoinhibitory adaptor that might be a useful target for immune suppression therapy.
Specifications
SAMSN1 | |
Polyclonal | |
Unconjugated | |
SAMSN1 | |
4930571B16Rik; 930571B16Rik; HACS1; hematopoietic adapter-containing SH3 and sterile α-motif (SAM) domains 1; hematopoietic adapter-containing SH3 and sterile alpha-motif (SAM) domains 1; hematopoietic adaptor containing SH3 and SAM domains 1; Nash; NASH1; nuclear localization signals, SAM and SH3 domain containing 1; SAM and SH3 domain containing 2; SAM domain, SH3 domain and nuclear localisation signals, 1; SAM domain, SH3 domain and nuclear localization signals 1; SAM domain, SH3 domain and nuclear localization signals protein 1; SAM domain, SH3 domain and nuclear localization signals, 1; SAM domain-containing protein SAMSN-1; SAMSN; Samsn1; SAMSN-1; SASH2; SH3 protein expressed in lymphocytes 2; SH3D6B; SH3-lymphocyte protein 2; SH3-SAM adaptor protein; SLy2; Src homology domain 3 (SH3)-containing adapter protein SH3 lymphocyte protein 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
170637, 64092, 67742 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P57725, Q9NSI8 | |
SAMSN1 | |
A synthesized peptide derived from human SAMSN1(Accession Q9NSI8), corresponding to amino acid residues S123-D173. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction