Alfa Aesar™ Sand, washed

Manufacturer: Alfa Aesar™ H379330B

Quantity 1000g

 View more versions of this product

Catalog No. AAH379330B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14808-60-7
    Linear Formula SiO2
    Formula Weight 60.08
    MDL Number MFCD02100519
    Chemical Name or Material Sand, Washed

    Specifications

    Formula Weight 60.08
    Health Hazard 3 GHS P Statement
    P260-P281-P308+P313-P314-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Get medical advice/attention if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H350-H373
    May cause cancer.
    May cause damage to organs through prolonged or repeated exposure.
    TSCA TSCA
    Odor Odorless
    Physical Form Granules
    MDL Number MFCD02100519
    Color Pale cream/pale brown
    RTECS Number W7330000
    Linear Formula SiO2
    Boiling Point 2230°C
    Quantity 1000g
    CAS 14808-60-7
    Melting Point 1710°C
    Health Hazard 1 Danger
    Chemical Name or Material Sand, Washed

    Description

    Applications
    Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H350-H373
    May cause cancer.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P281-P308+P313-P314-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Get medical advice/attention if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit