Learn More
Sand, washed
Description
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Boiling Point | 2230°C |
| Melting Point | 1710°C |
| Color | Brown-Cream |
| Physical Form | Granules |
| Quantity | 5000 g |
| Linear Formula | SiO2 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 60.08 |
| Odor | Odorless |
| Chemical Name or Material | Sand, Washed |
Safety and Handling
| Hazard Category | H350 |
| Hazard Statement | GHS H Statement H350-H373 May cause cancer. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 238-878-4 |
| RTECSNumber | W7330000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.