Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAND1 (Mona) Polyclonal Antibody, Abbomax

Catalog No. 502204556
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4556 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4556 Supplier Abbomax Inc Supplier No. 630140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Hela cells Cellular location: Cytoplasmic.

SAND1 (Mona) Polyclonal antibody specifically detects SAND1 (Mona) in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SAND1 (Mona)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene MON1A
Gene Accession No. Q6PDG8, Q86VX9
Gene Alias 2810468K17Rik; FLJ97088; MGC13272; MON1 homolog A; MON1 homolog A (yeast); MON1 homolog A, secretory trafficking associated; MON1 secretory trafficking family member A; Mon1a; RGD1308823; SAND1; Vacuolar fusion protein MON1 homolog A
Host Species Rabbit
Immunogen A synthetic peptide derived from a portion of the intra domain 190-240aa of human Mon1a protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72825, 84315
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.