Learn More
Eisco™ Sandstone Specimen
SCI_ED
Catalog No. S24739 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24739 12/Pk. 2 to 3 cm
S24738 1 Ea. 2 to 3 cm
S26665 10/Pk. 2 to 3 cm
S26682 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24739 Supplier Eisco™ Supplier No. ESNG0036PK12
Add to Cart
Add to Cart
Sandstone Specimen is made up of well-sorted and well-rounded grains of quartz.
  • For use in studying depositional environments
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Rounded, well-sorted sand grains are excellent identifiers for students during sedimentary rock identification
  • This indicates that clasts in rocks have traveled long distance from their source and were heavily eroded along way
Specifications
Product Type Sedimentary Rock
For Use With (Application) Studying depositional environments
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Sandstone