Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Sandstone Specimen
SCI_ED

Catalog No. S26682 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S26682 1 Ea. 7 to 8 cm
S24738 1 Ea. 2 to 3 cm
S26665 10/Pk. 2 to 3 cm
S24739 12/Pk. 2 to 3 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S26682 Supplier Eisco™ Supplier No. ESNG0036HAND
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sandstone Specimen is made up of well-sorted and well-rounded grains of quartz.

  • For use in studying depositional environments
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Rounded, well-sorted sand grains are excellent identifiers for students during sedimentary rock identification
  • This indicates that clasts in rocks have traveled long distance from their source and were heavily eroded along way

Specifications

Product Type Sedimentary Rock
Quantity 1 Ea.
Specimen Size 7 to 8 cm
Weight (Metric) 0.5 kg
Specimen Type Sandstone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.