Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Sanguinarine chloride
GREENER_CHOICE

Catalog No. 230225
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-022-5 25 mg
23-025 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-022-5 Supplier Tocris Bioscience™ Supplier No. 2302/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of protein phosphatase 2C (PP2C)

Sanguinarine chloride is an inhibitor of protein phosphatase 2C (PP2C), selective over PP1, PP2A and PP2B in vitro. Also inhibits mitogen-activated protein kinase phosphatase-1 (MKP-1) (IC50 = 10 μM). Plant alkaloid that exhibits anti-inflammatory and antioxidant activity. Blocks cell cycle in the G1 phase; inhibits cell growth in human prostate carcinoma cells.

Specifications

Chemical Name or Material 13-Methyl-[1,3]-benzodioxolo[5,6-c]-1,3-dioxolo[4,5-i]phenanthridinium chloride
CAS 5578-73-4
Assay Percent Range >97%
Quantity 25 mg
Molecular Formula C20H14ClNO4
Formula Weight Observed MW: 367.78
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.