Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Santacruzamate A
GREENER_CHOICESDP

Catalog No. 719150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-915-0 50 mg
71-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 71-915-0 Supplier Tocris Bioscience™ Supplier No. 7191/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent and selective HDAC2 inhibitor

Santacruzamate A is a highly potent and selective histone deactylase 2 (HDAC2) inhibitor (IC50 = 112 - 119 pM). Also inhibits HDAC6 in nanomolar range (IC50 = 433 nM). Displays >3500 fold selectivity for HDAC2 over HDAC6, and >8500-fold selectivity over HDAC4 (IC50 >1 μM for HDAC4). Inhibits growth of HuT-78 cutaneous T-cell lymphoma cell line. Identified as targeting human host proteins that interact with SARS-CoV-2.

Specifications

Chemical Name or Material Ethyl N-[4-oxo-4-[(2-phenylethyl)amino]butyl]carbamate
CAS 1477949-42-0
Quantity 50 mg
Target Class II Histone Deacetylases Inhibitors
Molecular Formula C15H22N2O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.