Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP-1 Rat anti-Mouse, Clone: 123, MilliporeSigma™

Catalog No. MABC1592MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1592MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1592MI Supplier MilliporeSigma Supplier No. MABC1592
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect SAP1 using this rat monoclonal Anti-SAP-1 Antibody, clone 123 Immunohistochemistry, Immunoprecipitation, and Western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP-1
Applications Immunohistochemistry, Western Blot, Immunoprecipitation, Immunofluorescence
Classification Monoclonal
Clone 123
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Ptprh,Sap-1
Host Species Rat
Immunogen Recombinant fusion protein comprising the extracellular region of mouse SAP-1 (amino acids 1-604) and the Fc region of human IgG.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 123 recognizes SAP-1 in HEK293A cells transfected with murine SAP-1. It targets an epitope in the extracellular domain.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.