Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SAP/SH2D1A, Rat, Unlabeled, Clone: 1A9,

Catalog No. MABF920MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF920MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF920MI Supplier MilliporeSigma™ Supplier No. MABF920
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SH2 domain-containing protein 1A (UniProt O60880; also known as Signaling lymphocytic activation molecule-associated protein, SLAM-associated protein, T-cell signal transduction molecule SAP) is encoded by the Sh2d1a (also known as Xlp) gene (Gene ID 20400) in murine species. SAP/SH2D1A is a 628-amino acid (isoform A) adaptor protein expressed in T cells, natural killer (NK) cells, NK-T cells, and some B cells. SAP/SH2D1A is made up almost exclusively of a Src homology 2 (SH2) domain (a.a. 6-104) and plays an important role in multiple immune cell functions, including T helper 2 (TH2) cytokine production, T cell-dependent antibody production, follicular T helper (TFH) cell differentiation, NK-T cell development, CD8+ T cell- and NK cell-mediated cytotoxicity. SAP couples signaling lymphocytic activation molecule (SLAM) family receptors to tyrosine phosphorylation signals through its interaction with the SH3 domain of Src family protein tyrosine kinase Fyn. In addition, SAP can prevent SH2 domain-containing negative regulators, such as phosphatases SHIP-1 and SHP-1, from inactivating SLAM-related receptors by blocking their interaction with the receptors. Evidence also shows that SAP/SH2D1A regulates T cell-dependent antibody production and TFH cell differentiation independent of Fyn. SAP/SH2D1A gene is mutated and inactivated in X-linked lymphoproliferative disease, a rare human immunodeficiency characterized by an uncontrolled immune response to Epstein-Barr virus infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP/SH2D1A
Applications Western Blot
Classification Monoclonal
Clone 1A9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Sh2d1a, Xlp
Gene Accession No. O88890
Host Species Rat
Immunogen GST-tagged recombinant full-length mouse SAP.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_035494
Test Specificity Clone 1A9 reacts with SAP, but not EAT-2 or ERT (Roncagalli, R., et al. (2005). Nat. Immunol. 6(10):1002-1010). Clone 1A9 immunostained wild-type, but not sh2d1a-knockout murine peripheral blood CD3+CD4+ lymphocytes (Zhong, M.C., and Veillette, A. (2013). Mol. Cell. Biol. 33(6):1223-1232).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.