Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAP (SLAM-Associated Protein) Monoclonal Antibody (XLP-1D12), PE, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Supplier:  Invitrogen™ 12978742

Encompass_Preferred

 View more versions of this product

Catalog No. 50-112-2460


Only null left
Add to Cart

Description

Description

Description: The monoclonal antibody XLP-1D12 reacts with human SLAM-associated protein (SAP), which is also known as SH2D1A. SAP is a 15-kDa SH2 domain-containing adaptor protein. Upon CD150/SLAM family receptor ligation, SAP recruits Fyn tyrosine kinase, which phosphorylates the receptor to activate downstream signaling events. SAP is expressed in thymocytes, T cells, NK cells, and NKT cells. Studies have demonstrated a role for SAP in promoting follicular T helper cell differentiation during germinal center formation. In addition, this protein is involved in NKT cell development, Th1/Th2 differentiation, development of innate T cells, and regulation of NK cell and CD8+ T cell cytotoxicity. Mutations of the gene encoding SAP results in X-linked lymphoproliferative (XLP) syndrome, which is characterized by impaired humoral immunity leading to increased susceptibility to Epstein-Barr virus infection and/or lymphoma. SAP has also been linked to automimmune diseases such as systemic lupus erythematosus. Applications Reported: This XLP-1D12 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This XLP-1D12 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transicription Factor Staining Buffer Set (cat. 00-5523) and protocol.

This gene encodes a protein that plays a major role in the bidirectional stimulation of T and B cells. This protein contains an SH2 domain and a short tail. It associates with the signaling lymphocyte-activation molecule, thereby acting as an inhibitor of this transmembrane protein by blocking the recruitment of the SH2-domain-containing signal-transduction molecule SHP-2 to its docking site. This protein can also bind to other related surface molecules that are expressed on activated T, B and NK cells, thereby modifying signal transduction pathways in these cells. Mutations in this gene cause lymphoproliferative syndrome X-linked type 1 or Duncan disease, a rare immunodeficiency characterized by extreme susceptibility to infection with Epstein-Barr virus, with symptoms including severe mononucleosis and malignant lymphoma. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SAP (SLAM-Associated Protein)
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
O60880
Sh2d1a
Affinity Chromatography
RUO
4068
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
XLP-1D12
PE
Sh2d1a
DSHP; Duncan disease homolog; Duncan disease SH2-protein; Duncan's disease; EBVS; FLJ18687; FLJ92177; Gm686; HGNC:10820; IMD5; LYP; MTCP1; RGD1562408; RP5-1052M9.3; SAP; SAP/SH2D1A; SH2 domain containing 1A; SH2 domain protein 1A; SH2 domain-containing protein 1A; SH2D1A; signaling lymphocyte activation molecule-associated protein; signaling lymphocytic activation molecule-associated protein; SLAM associated protein/SH2 domain protein 1A; SLAM-associated protein; SLAM-associated protein isoform SAP-2; T cell signal transduction molecule SAP; T-cell signal transduction molecule SAP; Xlp; XLPD; XLPD1
Rat
100 Tests
Primary
Human
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only.