Learn More
Invitrogen™ SAP102 Polyclonal Antibody

Description
PA1-045 detects synapse-associated protein 102 (SAP102) from rat tissues. PA1-045 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, this antibody detects an ~102 kDa protein representing SAP102 in rat brain homogenate. Immunohistochemical staining of SAP102 in rat hippocampus yields a pattern consistent with post-synaptic localization. The PA1-045 immunogen is a synthetic peptide corresponding to residues G(83) P V P P K P V P G K N T P K L N G(100) of rat SAP102. PA1-045 immunizing peptide (Cat. # PEP-023) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | SAP102 |
| Applications | Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | DLG3 |
| Gene Accession No. | Q62936 |
| Gene Alias | discs large homolog 3; discs large MAGUK scaffold protein 3; discs, large homolog 3; discs, large homolog 3 (Drosophila); disks large homolog 3; DLG3; Dlgh3; KIAA1232; mKIAA1232; MRX; MRX90; NEDLG; Neuroendocrine-DLG; PPP1R82; protein phosphatase 1, regulatory subunit 82; PSD-95/SAP90-related protein 1; SAP102; SAP-102; synapse-associated protein 102; XLMR |
| Gene Symbols | DLG3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.