Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAP130 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532007 Supplier Invitrogen™ Supplier No. PA532007
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (95%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SAP130 is a subunit of the histone deacetylase-dependent SIN3A corepressor complex (Fleischer et al., 2003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP130
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Sap130
Gene Accession No. Q8BIH0, Q9H0E3
Gene Alias 130 kDa Sin3-associated polypeptide; 2610304F09Rik; 6720406D06; FLJ12761; Histone deacetylase complex subunit SAP130; KIAA0017; mSin3A-associated protein 130; pre-mRNA splicing factor SF3b, 130 kDa subunit; pre-mRNA-splicing factor SF3b 130 kDa subunit; RGD1311657; RSE1; SAP 130; SAP130; SF3b130; SF3B3; Sin3A associated protein; Sin3A associated protein 130; Sin3A associated protein 130kDa; Sin3A associated protein, 130kDa; Sin3A-associated protein, 130kDa; Sin3-associated polypeptide p130; spliceosome-associated protein 130; Splicing factor 3B subunit 3; STAF130
Gene Symbols Sap130
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human SAP130. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269003, 79595
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.