Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAP18 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18938525UL
Description
SAP18 Polyclonal specifically detects SAP18 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SAP18 | |
Polyclonal | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
2HOR0202cell growth inhibiting protein 38, Cell growth-inhibiting gene 38 protein, cell growth-inhibiting protein 38, histone deacetlyase complex subunit SAP18, histone deacetylase complex subunit SAP18,18 kDa Sin3-associated polypeptide, MGC27131, SAP18P, Sin3A-associated protein, 18kDa, Sin3-associated polypeptide p18, sin3-associated polypeptide, 18 kDa, sin3-associated polypeptide, p18 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
SAP18 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VTQEEIKKEPEKPIDREKTCPLLLRVFTTNNGRHHRMDEFSRGNVPSSELQIYTWMDATLKELTSLVKEVYPEARKKGTHFNFAIVFTDVKRPGYRVKEIGSTMSGRKGTDDSMTLQSQKFQIGDYLDIAITPPNRAPP | |
25 μL | |
Stem Cell Markers | |
10284 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction