Learn More
Invitrogen™ SAP18 Monoclonal Antibody (3B2)

Description
Peptide Sequence: MAVESRVTQE EIKKEPEKPI DREKTCPLLL RVFTTNNGRH HRMDEFSRGN VPSSELQIYT WMDATLKELT SLVKEVYPEA RKKGTHFNFA IVFTDVKRPG YRVKEIGSTM SGRKGTDDSM TLQSQKFQIG DYLDIAITPP NRAPPTSGRM RPY.
Specifications
Specifications
| Antigen | SAP18 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3B2 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SAP18 |
| Gene Accession No. | O00422 |
| Gene Alias | 18 kDa Sin3-associated polypeptide; 2HOR0202; C530046K05Rik; cell growth inhibiting protein 38; cell growth-inhibiting gene 38 protein; cell growth-inhibiting protein 38; D11Ertd539e; EMegR4; GIG38; histone deacetlyase complex subunit SAP18; histone deacetylase complex subunit SAP18; SAP18; SAP18P; Sin3A associated protein 18; Sin3A associated protein 18kDa; Sin3A-associated protein, 18kDa; Sin3-associated polypeptide 18; sin3-associated polypeptide p18; sin3-associated polypeptide, 18 kDa; sin3-associated polypeptide, 18kDa; sin3-associated polypeptide, p18; Sinbp1 |
| Gene Symbols | SAP18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.