Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAP18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552821 Supplier Invitrogen™ Supplier No. PA552821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTQEEIKKEP EKPIDREKTC PLLLRVFTTN NGRHHRMDEF SRGNVPSSEL QIYTWMDATL KELTSLVKEV YPEARKKGTH FNFAIVFTDV KRPGYRVKEI GSTMSGRKGT DDSMTLQSQK FQIGDYLDIA ITPPNRAPP Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

SAP18 is involved in histone acetylation, which plays a key role in the regulation of eukaryotic gene expression. Histone acetylation and deacetylation are catalyzed by multisubunit complexes. The protein encoded by this gene is a component of the histone deacetylase complex, which includes SIN3, SAP30, HDAC1, HDAC2, RbAp46, RbAp48, and other polypeptides. This protein directly interacts with SIN3 and enhances SIN3-mediated transcriptional repression when tethered to the promoter. A pseudogene has been identified on chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP18
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAP18
Gene Accession No. O00422
Gene Alias 18 kDa Sin3-associated polypeptide; 2HOR0202; C530046K05Rik; cell growth inhibiting protein 38; cell growth-inhibiting gene 38 protein; cell growth-inhibiting protein 38; D11Ertd539e; EMegR4; GIG38; histone deacetlyase complex subunit SAP18; histone deacetylase complex subunit SAP18; SAP18; SAP18P; Sin3A associated protein 18; Sin3A associated protein 18kDa; Sin3A-associated protein, 18kDa; Sin3-associated polypeptide 18; sin3-associated polypeptide p18; sin3-associated polypeptide, 18 kDa; sin3-associated polypeptide, 18kDa; sin3-associated polypeptide, p18; Sinbp1
Gene Symbols SAP18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAP18. Recombinant protein control fragment (Product #RP-89364).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10284
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.