Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAP25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564036 Supplier Invitrogen™ Supplier No. PA564036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGHWNGQQAP PDAGFPVVCC EDVFLSDPLL PRGQRVPLYL SKAPQQMMGS L Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 57%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP25
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAP25
Gene Accession No. Q8TEE9
Gene Alias 25 kDa Sin3-associated polypeptide; Histone deacetylase complex subunit SAP25; SAP25; Sin3 corepressor complex subunit SAP25; Sin3A associated protein 25; Sin3A associated protein 25kDa; Sin3A-associated protein, 25kDa; sin3A-binding protein 25; sin3-associated polypeptide, 25kDa
Gene Symbols SAP25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAP25. Recombinant protein control fragment (Product #RP-101992).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100316904
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.