Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAP30 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299031JF549
Description
SAP30 Polyclonal specifically detects SAP30 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SAP30 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
30 kDa Sin3-associated polypeptide, histone deacetylase complex subunit SAP30, Sin3 corepressor complex subunit SAP30, Sin3A-associated protein, 30kDa, Sin3-associated polypeptide p30, sin3-associated polypeptide, 30 kDa, Sin3-associated polypeptide, 30kDa | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human SAP30. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
8819 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction