Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAP30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566229 Supplier Invitrogen™ Supplier No. PA566229
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEKDTLTYFIY SVKNDKNKSD LKVDSGVH Highest antigen sequence identity to the following orthologs - mouse 90%, rat 90%.

Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP30
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAP30
Gene Accession No. O75446
Gene Alias 30 kDa Sin3-associated polypeptide; 30kDa; histone deacetylase complex subunit SAP30; Sap30; sin3 associated polypeptide; sin3 associated polypeptide, 30kD; sin3 corepressor complex subunit SAP30; Sin3A associated protein 30; Sin3A associated protein 30kDa; Sin3A-associated protein; Sin3A-associated protein, 30kDa; Sin3-associated polypeptide p30; sin3-associated polypeptide, 30 kDa; Sin3-associated polypeptide, 30kDa; Sin3-associated protein
Gene Symbols SAP30
Host Species Rabbit
Immunogen Recombinant Human SAP30. Recombinant protein control fragment (Product #RP-106908).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8819
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.