Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAP30BP Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP147298H
Description
SAP30BP Polyclonal antibody specifically detects SAP30BP in Human, Mouse samples. It is validated for Western BlotSpecifications
SAP30BP | |
Polyclonal | |
Western Blot | |
HCNGPDKFZp586L2022, HTRGHSV-1 binding, HTRPSAP30-binding protein, SAP30 binding protein, Transcriptional regulator protein HCNGP | |
The immunogen for this product maps to a region between residue 90 and 140 of human SAP30 Binding Protein using the numbering given in entry NP_037392.1 (GeneID 29115). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
29115 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction