Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAP30BP Antibody (OTI4H1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273987AF594
Description
SAP30BP Monoclonal specifically detects SAP30BP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SAP30BP | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
SAP30BP | |
Recombinant protein fragment corresponding to amino acids 1-260 of human SAP30BP (NP_037392) produced in E.coli. | |
0.1 mL | |
29115.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4H1 | |
Western Blot, Immunohistochemistry | |
HCNGPDKFZp586L2022, HTRGHSV-1 binding, HTRPSAP30-binding protein, SAP30 binding protein, Transcriptional regulator protein HCNGP | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction