Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAP30BP Monoclonal Antibody (OTI4H1)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526587

Catalog No. PIMA526587


Only null left
Add to Cart

Description

Description

SAP30BP Monoclonal Antibody for Western Blot, IHC (P)

Induces cell death. May act as a transcriptional corepressor of a gene related to cell survival. May be involved in the regulation of beta-2-microglobulin genes. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SAP30BP
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UHR5
Sap30bp
Human recombinant protein fragment corresponding to amino acids 1-260 of SAP30BP produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI4H1
Unconjugated
Sap30bp
2700016D05Rik; fb33d03; Hcngp; HSV-1 binding; HTRG; HTRP; RGD1565537; SAP30 binding protein; SAP30-binding protein; Sap30bp; transcriptional regulator protein; transcriptional regulator protein (Hcngp); transcriptional regulator protein HCNGP; Unknown (protein for MGC:142452); wu:fb33d03; zgc:55340
Mouse
Affinity Chromatography
RUO
29115
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.