Learn More
SAP30BP Mouse anti-Human, Clone: OTI4A9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803084

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Induces cell death. May act as a transcriptional corepressor of a gene related to cell survival. May be involved in the regulation of beta-2-microglobulin genes.Specifications
SAP30BP | |
Monoclonal | |
Unconjugated | |
SAP30BP | |
HCNGP, HTRG, HTRP | |
Mouse | |
Affinity Chromatography | |
RUO | |
29115 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UHR5 | |
SAP30BP | |
Human recombit protein fragment corresponding to amino acids 1-260 of human SAP30BP produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.