Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAP30BP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111152
Description
Immunogen sequence: SEDSYYEALA KAQKIEMDKL EKAKKERTKI EFVTGTKKGT TTNATSTTTT TASTAVADAQ KRKSKWDSAI PVTTIAQPTI LTTTATLPAV VTVTTSASGS KTTVISAVGT.
Induces cell death. May act as a transcriptional corepressor of a gene related to cell survival. May be involved in the regulation of beta-2-microglobulin genes. [UniProt].
Specifications
SAP30BP | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UHR5 | |
Sap30bp | |
Recombinant Protein corresonding to Human SAP30BP. Recombinant protein control fragment (Product #RP-91680). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Sap30bp | |
2700016D05Rik; fb33d03; Hcngp; HSV-1 binding; HTRG; HTRP; RGD1565537; SAP30 binding protein; SAP30-binding protein; Sap30bp; transcriptional regulator protein; transcriptional regulator protein (Hcngp); transcriptional regulator protein HCNGP; Unknown (protein for MGC:142452); wu:fb33d03; zgc:55340 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29115 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction