Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAP30BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529431
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529431 Supplier Invitrogen™ Supplier No. PA529431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Molt-4. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Induces cell death. May act as a transcriptional corepressor of a gene related to cell survival. May be involved in the regulation of beta-2-microglobulin genes. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP30BP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Sap30bp
Gene Accession No. Q9UHR5
Gene Alias 2700016D05Rik; fb33d03; Hcngp; HSV-1 binding; HTRG; HTRP; RGD1565537; SAP30 binding protein; SAP30-binding protein; Sap30bp; transcriptional regulator protein; transcriptional regulator protein (Hcngp); transcriptional regulator protein HCNGP; Unknown (protein for MGC:142452); wu:fb33d03; zgc:55340
Gene Symbols Sap30bp
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human SAP30BP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29115
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.