Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAP30L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA146138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA146138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA146138 Supplier Invitrogen™ Supplier No. PA146138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody does not react with endogenous non-human SAP30L. Immunogen sequence: VKSNKSRLDQ KSEGGKQLE.

A novel TGF- beta up-regulated mRNA species, the Sin3-associated protein 30-like, SAP30L has been identified. The predicted nuclear localization signal of SAP30L is sufficient for nuclear transport of the protein although mutating it does not completely remove SAP30L from the nuclei. By reason of its nuclear localization and close homology to SAP30 it is thought that SAP30L might have a role in recruiting the Sin3-histone deacetylase complex to specific co-repressor complexes in response to TGF-beta, leading to the silencing of proliferation-driving genes in the differentiating intestinal epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP30L
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SAP30L
Gene Accession No. Q9HAJ7
Gene Alias 2310079P12Rik; AF006998; DKFZp667L2214; FLJ11526; FLJ23595; FLJ36497; HCV non-structural protein 4A-transactivated protein 2; Histone deacetylase complex subunit SAP30L; L55; NS4ATP2; SAP30 like; SAP30L; SAP30-like; Sin3 corepressor complex subunit SAP30L; Sin3A associated protein p30-like; sin3A-associated protein p30-like protein; sin3-associated protein p30-like
Gene Symbols SAP30L
Host Species Rabbit
Immunogen A synthetic peptide made to residues 165-183 of human SAP30L.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79685
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.