Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAP30L Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142670
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
A novel TGF- beta up-regulated mRNA species, the Sin3-associated protein 30-like, SAP30L has been identified. The predicted nuclear localization signal of SAP30L is sufficient for nuclear transport of the protein although mutating it does not completely remove SAP30L from the nuclei. By reason of its nuclear localization and close homology to SAP30 it is thought that SAP30L might have a role in recruiting the Sin3-histone deacetylase complex to specific co-repressor complexes in response to TGF-beta, leading to the silencing of proliferation-driving genes in the differentiating intestinal epithelial cells.
Specifications
| SAP30L | |
| Polyclonal | |
| Unconjugated | |
| SAP30L | |
| 2310079P12Rik; AF006998; DKFZp667L2214; FLJ11526; FLJ23595; FLJ36497; HCV non-structural protein 4A-transactivated protein 2; Histone deacetylase complex subunit SAP30L; L55; NS4ATP2; SAP30 like; SAP30L; SAP30-like; Sin3 corepressor complex subunit SAP30L; Sin3A associated protein p30-like; sin3A-associated protein p30-like protein; sin3-associated protein p30-like | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 360531 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| 0 | |
| SAP30L | |
| Peptide with sequence C-RLDQKSEGSKQLE (aa 170-182). | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction