Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112884
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112884 Supplier Invitrogen™ Supplier No. PA5112884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SAP9 Polyclonal Antibody for Western Blot, ELISA

Secreted aspartic peptidases (SAPs) are a group of ten acidic hydrolases considered as key virulence factors. These enzymes supply the fungus with nutrient amino acids as well as are able to degrade the selected host's proteins involved in the immune defense. Involved in triggering host polymorphonuclear neutrophils chemotaxis toward germ tubes. Moreover, acts toward human hemoglobin though limited proteolysis to generate a variety of antimicrobial hemocidins, enabling to compete with the other microorganisms of the same physiological niche using the microbicidal peptides generated from the host protein. Required for cell surface integrity and cell separation during budding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SAP9
Gene Accession No. Q59SU1
Gene Symbols SAP9
Host Species Rabbit
Immunogen Recombinant Yeast Candidapepsin-9 protein (18-544AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3645625
Target Species Yeast
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.