Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAP97 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585827 Supplier Invitrogen™ Supplier No. PA585827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Human (89%), Mouse (100%), Rat (90%), Rhesus Monkey (89%). Positive Control: mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP97
Applications Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene DLG1
Gene Accession No. Q62696, Q811D0
Gene Alias B130052P05Rik; discs large homolog 1; discs large homolog 1, scribble cell polarity complex component; discs large MAGUK scaffold protein 1; discs, large homolog 1; discs, large homolog 1 (Drosophila); discs, large homolog 1 (Drosophila) transcript variant 6-v1; discs, large homolog 1 (Drosophila) transcript variant 6-v2; disks large homolog 1; dJ1061C18.1.1; DKFZp761P0818; DKFZp781B0426; DLG1; Dlgh1; Drosophila discs-large tumor suppressor homologue (synapse associated protein); E-dlg/SAP97; Embryo-dlg/synapse-associated protein 97; hDlg; mKIAA4187; presynaptic protein SAP97; SAP97; SAP-97; synapse-associated protein 97
Gene Symbols DLG1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of mouse SAP97. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13383, 25252
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.