Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAPAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578658
Description
Positive Control: mouse hippocampus, mouse brain, rat brain Predicted Reactivity: Human (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Part of the postsynaptic scaffold in neuronal cells. Target binds the guanylate kinase-like domain of the PSD-95 family of synaptic ion channel clustering proteins; may play a role in protein-protein interactions at the synaptic junction.
Specifications
SAPAP1 | |
Polyclonal | |
Unconjugated | |
Dlgap1 | |
4933422O14Rik; 9630002F18; AI845682; AI848168; BB075781; D17Bwg0511e; DAP1; DAP-1; DAP-1 beta; DAP-1-ALPHA; DAP-1-BETA; discs large homolog associated protein 1; discs, large (Drosophila) homolog-associated protein 1; disks large-associated protein 1; DLG associated protein 1; Dlgap1; DLGAP1A; DLGAP1B; GKAP; GKAP/SAPAP; GKAP/SAPAP1; GKPA/SAPAP; guanylate kinase associated protein; guanylate kinase-associated protein; hGKAP; Kiaa4162; mKIAA4162; PSD-95 binding protein; PSD-95/SAP90 binding protein 1; PSD-95/SAP90-binding protein 1; rGKAP; SAP90/PSD-95 associated protein 1; SAP90/PSD-95-associated protein 1; SAPAP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
224997, 65040 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.87 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P97836, Q9D415 | |
Dlgap1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse DLGAP1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction