Learn More
Invitrogen™ SAPAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588953
Description
Immunogen sequence: MKGLSGSRSH HHGVTCDSAC DSLSHHSDRK PYLLSPVEHH PADHPYYTQR NSFQAECVGP FSDPLASSTF PRRHYTSQQE LKDECALVPR TLATKANRIP ANLLDQFERQ LPLSRDGYHT LQYKRTAVEH RSDSPGRIRH LVHSVQKLFT KSHSLEGPSK GSVNGGKASP DEAQAARYGK RSKSKERRAE PKARPSTSPG WWSSDDNLDG DMCIYHAPSG VMTMGRCPDR SASQYFLEAY NTISEQAVKA; Positive Samples: U-87MG, Mouse brain; Cellular Location: Cell junction, Cell membrane, Peripheral membrane protein, postsynaptic cell membrane, postsynaptic density, synapse.
Part of the postsynaptic scaffold in neuronal cells. Target binds the guanylate kinase-like domain of the PSD-95 family of synaptic ion channel clustering proteins; may play a role in protein-protein interactions at the synaptic junction.
Specifications
SAPAP1 | |
Polyclonal | |
Unconjugated | |
Dlgap1 | |
4933422O14Rik; 9630002F18; AI845682; AI848168; BB075781; D17Bwg0511e; DAP1; DAP-1; DAP-1 beta; DAP-1-ALPHA; DAP-1-BETA; discs large homolog associated protein 1; discs, large (Drosophila) homolog-associated protein 1; disks large-associated protein 1; DLG associated protein 1; Dlgap1; DLGAP1A; DLGAP1B; GKAP; GKAP/SAPAP; GKAP/SAPAP1; GKPA/SAPAP; guanylate kinase associated protein; guanylate kinase-associated protein; hGKAP; Kiaa4162; mKIAA4162; PSD-95 binding protein; PSD-95/SAP90 binding protein 1; PSD-95/SAP90-binding protein 1; rGKAP; SAP90/PSD-95 associated protein 1; SAP90/PSD-95-associated protein 1; SAPAP1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
224997, 9229 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.58 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14490, Q9D415 | |
Dlgap1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human DLGAP1 (NP_0012296901). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.