Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAPAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119751
This item is not returnable.
View return policy
Description
Positive Control: Human brain tissue cell lysates, PC-12, N2A, SHG-44, rat testis tissue, rat brain tissue, mouse testis tissue, mouse brain tissue, Daudi. Subcellular Location: Cell junction, Cell membrane, Membrane, Synapse.
Part of the postsynaptic scaffold in neuronal cells. Target binds the guanylate kinase-like domain of the PSD-95 family of synaptic ion channel clustering proteins; may play a role in protein-protein interactions at the synaptic junction.
Specifications
SAPAP1 | |
Polyclonal | |
Unconjugated | |
Dlgap1 | |
4933422O14Rik; 9630002F18; AI845682; AI848168; BB075781; D17Bwg0511e; DAP1; DAP-1; DAP-1 beta; DAP-1-ALPHA; DAP-1-BETA; discs large homolog associated protein 1; discs, large (Drosophila) homolog-associated protein 1; disks large-associated protein 1; DLG associated protein 1; Dlgap1; DLGAP1A; DLGAP1B; GKAP; GKAP/SAPAP; GKAP/SAPAP1; GKPA/SAPAP; guanylate kinase associated protein; guanylate kinase-associated protein; hGKAP; Kiaa4162; mKIAA4162; PSD-95 binding protein; PSD-95/SAP90 binding protein 1; PSD-95/SAP90-binding protein 1; rGKAP; SAP90/PSD-95 associated protein 1; SAP90/PSD-95-associated protein 1; SAPAP1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
224997, 65040, 9229 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
O14490, P97836, Q9D415 | |
Dlgap1 | |
Synthetic peptide within human GKAP aa 380-430. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction