Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAPAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555312
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555312 Supplier Invitrogen™ Supplier No. PA555312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSDSGFLAGG RPPGEPGGPF CLEGPDGSYR DLSFKGRSGG SEGRCLACTG MSMSLDGQSV KRSAWHTMMV SQGRDGYPGA GPGKGLLGPE TKAKARTYHY LQVPQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

SAP90/PSD-95-associated protein 3 (SAPAP3, also known as DLGAP3) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP3 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). Both SAPAP3 protein and mRNA are targeted to dendrites, whereas SAPAP1, -2, and -4 mRNAs are detected mainly in cell bodies. Recent experiments have suggested that SAPAP3 may be involved in obsessive-compulsive disorder (OCD), as mice lacking SAPAP3 exhibited OCD-like symptoms which could be relieved by lentiviral-mediated selective expression of SAPAP3 in the striatum of SAPAP3-mutant mice. At least two isoforms are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPAP3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DLGAP3
Gene Accession No. O95886
Gene Alias BC058433; DAP3; DAP-3; discs large homolog associated protein 3; discs, large (Drosophila) homolog-associated protein 3; disks large-associated protein 3; DLG associated protein 3; Dlgap3; Prpl8; PSD-95/SAP90-associated protein-3; PSD-95/SAP90-binding protein 3; SAP90/PSD 95 associated protein 3; SAP90/PSD-95-associated protein 3; SAPAP3
Gene Symbols DLGAP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAPAP3. Recombinant protein control fragment (Product #RP-94082).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58512
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.