Learn More
Invitrogen™ SAPAP3 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PKTSPKAVAR RFTTRRSSSV DQARINCCVP PRIHPRSSIP GYSRSLTTGQ LSDELNQQLE AVCGSVFGEL ES Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 92%.
Specifications
Specifications
| Antigen | SAPAP3 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DLGAP3 |
| Gene Accession No. | O95886 |
| Gene Alias | BC058433; DAP3; DAP-3; discs large homolog associated protein 3; discs, large (Drosophila) homolog-associated protein 3; disks large-associated protein 3; DLG associated protein 3; Dlgap3; Prpl8; PSD-95/SAP90-associated protein-3; PSD-95/SAP90-binding protein 3; SAP90/PSD 95 associated protein 3; SAP90/PSD-95-associated protein 3; SAPAP3 |
| Gene Symbols | DLGAP3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.