Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAPAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562919 Supplier Invitrogen™ Supplier No. PA562919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVQVEDDWRS SVPSHSMSSR RDTDSDTQDA NDSSCKSSER SLPDCTPHPN SISIDAGPRQ APKIAQIKRN LSYGDNSDPA L Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 98%.

SAP90/PSD-95-associated protein 4 (SAPAP4, also known as DLGAP4) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP4 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). SAPAP4 mRNA is targeted to cell bodies in a similar manner to SAPAP1 and -2, whereas SAPAP3 mRNA is detected mainly in cell bodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPAP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DLGAP4
Gene Accession No. Q9Y2H0
Gene Alias AI225853; BC024558; Dap4; DAP-4; discs large homolog associated protein 4; discs, large (Drosophila) homolog-associated protein 4; discs, large homolog-associated protein 4; discs, large homolog-associated protein 4 (Drosophila); disks large-associated protein 4; DLG associated protein 4; Dlgap4; DLP4; Kiaa0964; PSD-95/SAP90-associated protein-4; PSD-95/SAP90-binding protein 4; RP5-977B1.6; SAP90/PSD-95-associated protein 4; Sapap4; SAPAP-4; WBP16; WW domain-binding protein 16
Gene Symbols DLGAP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAPAP4. Recombinant protein control fragment (Product #RP-99293).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.