Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAPAP4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520558

Catalog No. PIPA520558


Only null left
Add to Cart

Description

Description

A suggested positive control is SK-N-SH cell lysate. PA5-20558 can be used with blocking peptide PEP-0678.

SAP90/PSD-95-associated protein 4 (SAPAP4, also known as DLGAP4) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP4 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). SAPAP4 mRNA is targeted to cell bodies in a similar manner to SAPAP1 and -2, whereas SAPAP3 mRNA is detected mainly in cell bodies.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SAPAP4
Polyclonal
Unconjugated
DLGAP4
AI225853; BC024558; Dap4; DAP-4; discs large homolog associated protein 4; discs, large (Drosophila) homolog-associated protein 4; discs, large homolog-associated protein 4; discs, large homolog-associated protein 4 (Drosophila); disks large-associated protein 4; DLG associated protein 4; Dlgap4; DLP4; Kiaa0964; PSD-95/SAP90-associated protein-4; PSD-95/SAP90-binding protein 4; RP5-977B1.6; SAP90/PSD-95-associated protein 4; Sapap4; SAPAP-4; WBP16; WW domain-binding protein 16
Rabbit
Antigen affinity chromatography
RUO
22839
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9Y2H0
DLGAP4
A 13 amino acid peptide from near the center of human SAPAP4.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.