Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAPCD1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984313
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4313 Supplier Bioss Supplier No. BS19387R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SAPCD1 is a protein coding gene.

Specifications

Antigen SAPCD1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SAPCD1
Gene Accession No. Q5SSQ6
Gene Alias BOS_21652; C23H6orf26; C6orf26; G7D; NG23; Protein G7d; SAPCD1; suppressor APC domain containing 1; Suppressor APC domain-containing protein 1
Gene Symbols SAPCD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SAPCD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401251
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.